Merck
  • Feasibility of using algorithm-based clinical decision support for symptom assessment and management in lung cancer.
Feasibility of using algorithm-based clinical decision support for symptom assessment and management in lung cancer.
Journal of pain and symptom management (2014-06-01)
Mary E Cooley, Traci M Blonquist, Paul J Catalano, David F Lobach, Barbara Halpenny, Ruth McCorkle, Ellis B Johns, Ilana M Braun, Michael S Rabin, Fatma Zohra Mataoui, Kathleen Finn, Donna L Berry, Janet L Abrahm
ABSTRACT
Distressing symptoms interfere with the quality of life in patients with lung cancer. Algorithm-based clinical decision support (CDS) to improve evidence-based management of isolated symptoms seems promising, but no reports yet address multiple symptoms. This study examined the feasibility of CDS for a Symptom Assessment and Management Intervention targeting common symptoms in patients with lung cancer (SAMI-L) in ambulatory oncology. The study objectives were to evaluate completion and delivery rates of the SAMI-L report and clinician adherence to the algorithm-based recommendations. Patients completed a web-based symptom assessment and SAMI-L created tailored recommendations for symptom management. Completion of assessments and delivery of reports were recorded. Medical record review assessed clinician adherence to recommendations. Feasibility was defined as 75% or higher report completion and delivery rates and 80% or higher clinician adherence to recommendations. Descriptive statistics and generalized estimating equations were used for data analyses. Symptom assessment completion was 84% (95% CI=81-87%). Delivery of completed reports was 90% (95% CI=86-93%). Depression (36%), pain (30%), and fatigue (18%) occurred most frequently, followed by anxiety (11%) and dyspnea (6%). On average, overall recommendation adherence was 57% (95% CI=52-62%) and was not dependent on the number of recommendations (P=0.45). Adherence was higher for anxiety (66%; 95% CI=55-77%), depression (64%; 95% CI=56-71%), pain (62%; 95% CI=52-72%), and dyspnea (51%; 95% CI=38-64%) than for fatigue (38%; 95% CI=28-47%). The CDS systems, such as SAMI-L, have the potential to fill a gap in promoting evidence-based care.