Merck
  • A reduced-toxicity regimen is associated with durable engraftment and clinical cure of nonmalignant genetic diseases among children undergoing blood and marrow transplantation with an HLA-matched related donor.
A reduced-toxicity regimen is associated with durable engraftment and clinical cure of nonmalignant genetic diseases among children undergoing blood and marrow transplantation with an HLA-matched related donor.
Biology of blood and marrow transplantation : journal of the American Society for Blood and Marrow Transplantation (2014-12-03)
Kris Michael Mahadeo, Kenneth I Weinberg, Hisham Abdel-Azim, David B Miklos, Renna Killen, Donald Kohn, Gay M Crooks, Ami J Shah, Sandhya Kharbanda, Rajni Agarwal, Neena Kapoor
ABSTRACT
Blood and marrow transplantation (BMT) is a standard curative therapy for patients with nonmalignant genetic diseases. Myeloablative conditioning has been associated with significant regimen-related toxicity (RRT), whereas reduced-intensity conditioning regimens have been associated with graft failure. In this prospective pilot trial conducted at 2 centers between 2006 and 2013, we report the outcome of 22 patients with nonmalignant genetic diseases who were conditioned with a novel reduced-toxicity regimen: i.v. busulfan (16 mg/kg), alemtuzumab (52 mg/m(2)), fludarabine (140 mg/m(2)), and cyclophosphamide (105 mg/kg). The median age of the study population was 3.5 years (range, 5 months to 26 years). No cases of sinusoidal obstruction syndrome, severe or chronic graft-versus-host disease (GVHD), or primary graft failure were reported. Median time to neutrophil engraftment (>500 cells/μL) and platelet engraftment (>20K cells/μL) were 19 (range, 12 to 50) and 23.5 (range, 14 to 134) days, respectively. The median length of follow-up was 3 years (range, .2 to 6.3). The overall survival rates were 95% at 100 days (95% confidence interval, .72 to .99) and 90% at 6 years (95% confidence interval, .68 to .98). RRT and chronic GVHD are significant barriers to BMT for patients with nonmalignant genetic diseases. This alemtuzumab-based reduced-toxicity regimen appears to be promising with durable engraftment, effective cure of clinical disease, low rates of RRT, and no observed chronic GVHD.
Product Description
Busulfan, European Pharmacopoeia (EP) Reference Standard