- Experimental and theoretical studies on the hydrogen-bond-promoted enantioselective hetero-Diels-alder reaction of Danishefsky's diene with benzaldehyde.
Experimental and theoretical studies on the hydrogen-bond-promoted enantioselective hetero-Diels-alder reaction of Danishefsky's diene with benzaldehyde.
The Journal of Organic Chemistry (2006)
Zhang X, et al.