Merck
  • REG3A accelerates pancreatic cancer cell growth under IL-6-associated inflammatory condition: Involvement of a REG3A-JAK2/STAT3 positive feedback loop.
REG3A accelerates pancreatic cancer cell growth under IL-6-associated inflammatory condition: Involvement of a REG3A-JAK2/STAT3 positive feedback loop.
Cancer letters (2015-03-18)
Xiulan Liu, Jun Wang, Hongjie Wang, Guoxiao Yin, Yang Liu, Xiang Lei, Ming Xiang
ABSTRACT
Regenerating gene protein (REG) 3A is a 19 kD secretory pancreas protein with pro-growth function. Previously we demonstrated that overexpression of REG3A, acting as a key molecule for up-regulation of the JAK2/STAT3 pathway, contributed to inflammation-related pancreatic cancer (PaC) development. However the exact network associated with REG3A signaling still remains unclear. Here we determined that exposure of human PaC cells to cytokine IL-6 activated the oncogenic JAK2/STAT3 pathway, which directly upregulated REG3A expression, accelerated cell cycle progression by promoting CyclinD1 expression, and enhancing the expression of the anti-apoptosis Bcl family. Importantly, the activation of REG3A would instead enhance the JAK2/STAT3 pathway to constitute a REG3A-JAK2/STAT3 positive feedback loop, which leads to the amplification of the oncogenic effects of IL-6/JAK2/STAT3, a classic pathway linking to inflammation-related tumorigenesis, ultimately resulting in PaC cell over-proliferation and tumor formation both in vitro and in vivo. Moreover, EGFR was found to mediate the REG3A signal for PaC cell growth and JAK2/STAT3 activation, thus functioning as a REG3A receptor. Collectively, our results provide the first evidence for the presence of the synergistic effect of REG3A and IL-6 on PaC development via a REG3A-JAK2/STAT3 positive feedback loop.