Merck
  • 1,25-Dihydroxyvitamin D3 alleviates salivary adenoid cystic carcinoma progression by suppressing GPX1 expression through the NF-κB pathway.
1,25-Dihydroxyvitamin D3 alleviates salivary adenoid cystic carcinoma progression by suppressing GPX1 expression through the NF-κB pathway.
International journal of oncology (2016-01-20)
Zhiquan Huang, Yeqing Liu, Zixian Huang, Haifeng Li, Xiangfeng Gan, Zhuojian Shen
ABSTRACT
1,25-Dihydroxyvitamin D3 (1,25D3) is the active form of vitamin D with antineoplastic effects. The glutathione peroxidase-1 (GPX1) gene is associated with tumour progression. The present study aimed to explore the role of GPX1 in 1,25D3-mediated progression of salivary adenoid cystic carcinoma (SACC). Downregulating GPX1 expression inhibited SACC cell proliferation, chemoresistance, motility, and uPA secretion, but promoted apoptosis via the NF-κB pathway. Pre-processing 1,25D3 inhibited expression of NF-κB/GPX1/uPA, which subsequently suppressed cell motility and cisplatin-resistance in ACC-2 cells. In conclusion, 1,25D3 works as a modifier of NF-κB/GPX1/uPA expression, inhibiting cisplatin-resistance and cell invasive ability of SACC cells. The present study comprehensively elucidated the potential mechanism underlying the effects of vitamin D on chemoresistance and invasive potential in SACC.