Merck
  • Knockdown of WISP1 inhibit proliferation and induce apoptosis in ALL Jurkat cells.
Knockdown of WISP1 inhibit proliferation and induce apoptosis in ALL Jurkat cells.
International journal of clinical and experimental pathology (2016-01-30)
Xiaomin Zhang, Xi Chen, Juan Liu, Xiushuai Dong, Yinglan Jin, Yaoyao Tian, Yanming Xue, Liyan Chen, Yuying Chang, Yao Liu, Jinghua Wang
ABSTRACT
WISP1, a Wnt-induced secreted protein, has been found to have anticancer activity. ALL is a leading cause of death. Here we investigate the WISP1 effects on ALL Jurkat cells. Cell viability was assessed by CCK-8. Cell cycle and apoptosis were detected by flow cytometry. Mitochondrial membrane potential (MMP) was monitored using TMRM. Generation of reactive oxygen species (ROS) was quantified using DCFH-DA. Western blot was used to detect the expression of cell proliferation and apoptosis related genes. The results showed that knockdown of WISP1 significantly inhibited proliferation of Jurkat cells. Parallelly, cell cycle distribution was increased at G1 phase and apoptotic rate was induced after WISP1 knockdown. Furthermore, knockdown of WISP1 induced apoptosis of Jurkat cells was also associated with loss of MMP and generation of ROS. Western blot results showed that the protein expression p-AKT, PCNA, CDK1, P-ERK, CDK2, VEGF, VEGFR2 and Bcl2 were decreased, while the expression of Bax was up-regulated. In conclusion, WISP1 plays an important role in proliferation and apoptosis of Jurkat cells in mitochondria dependent pathway, the specific mechanisms need further study.
Sigma-Aldrich
Tetramethylrhodamine methyl ester perchlorate, ≥95%