Merck
  • Chemical composition and antimicrobial and antioxidant activities of the essential oil and methanol extract of Hippomarathrum microcarpum (Bieb.) from Turkey.
Chemical composition and antimicrobial and antioxidant activities of the essential oil and methanol extract of Hippomarathrum microcarpum (Bieb.) from Turkey.
Journal of agricultural and food chemistry (2007-02-01)
Hakan Ozer, Münevver Sökmen, Medine Güllüce, Ahmet Adigüzel, Fikrettin Sahin, Atalay Sökmen, Hamdullah Kiliç, Ozlem Baris
ABSTRACT
Hippomarathrum microcarpum grows wild in eastern Anatolia, Turkey, and is a plant utilized as food by people. In this study, the in vitro antimicrobial and antioxidant activities of the essential oil and methanol extract from H. microcarpum and its essential oil composition were investigated. The essential oil, which has bornyl acetate, caryophyllene oxide, and beta-caryophyllene as its main components, exhibited activity against eight bacteria, nine fungi, and a yeast, Candida albicans, with minimum inhibitory concentration values ranging from 62.50 to 125 muL/mL; the methanol extract showed weak activity. The antioxidant activity of these extracts was assessed by the beta-carotene bleaching test and the 1,1'-diphenyl-2-picrylhydrazyl (DPPH) radical scavenging test. The inhibition of linoleic acid oxidation was very weak for both extracts tested. The inhibition percentages were found to be 22.9 and 33.5% for methanol and essential oil, respectively, at the concentration of 2 g/L. The oil scavenged DPPH at higher concentrations (IC50 = 10.69 +/- 0.05 mg/mL), but the methanol extract exhibited no activity. The total phenolic content of the methanol extract was found to be 4.7 +/- 0.1%.
Sigma-Aldrich
Amphotericin B from Streptomyces sp., ~80% (HPLC), powder
Pricing and availability is not currently available.