- Synthesis, structural and spectroscopic studies of 2-oxoacenaphthylen-1(2H)-ylidene nicotinohydrazide.
Synthesis, structural and spectroscopic studies of 2-oxoacenaphthylen-1(2H)-ylidene nicotinohydrazide.
Spectrochimica acta. Part A, Molecular and biomolecular spectroscopy (2016-03-06)
M S C Henriques, R Del Amparo, D Pérez-Álvarez, B A Nogueira, M C Rodríguez-Argüelles, J A Paixão
PMID26944509
ABSTRACT
The synthesis of a new hydrazone, 2-oxoacenaphthylen-1(2H)-ylidene nicotinohydrazide, and its structural and spectroscopic characterization is reported. The obtained powder was recrystallized from DMSO and ethanol that afforded small crystals used for single-crystal X-ray diffraction studies. The compound was found to crystallize in two polymorphs, depending on the crystallization conditions. One of the polymorphs (form I) crystallizes in the centrosymmetric P2