Merck
  • Overexpression of OCT4 is associated with gefitinib resistance in non-small cell lung cancer.
Overexpression of OCT4 is associated with gefitinib resistance in non-small cell lung cancer.
Oncotarget (2016-11-07)
Bin Li, Zhouhong Yao, Yunyan Wan, Dianjie Lin
ABSTRACT
Epidermal growth factor receptor (EGFR)-targeted tyrosine kinase inhibitors (TKIs) have emerged as first-line drugs for non-small cell lung cancers (NSCLCs). However, the resistance to TKIs represents the key limitation for their therapeutic efficacy. We found that the difference of OCT4 expression between NSCLC and the adjacent non-tumourous tissues was statistically significant. Knockdown of OCT4 in NSCLC cells could decrease cell proliferation, and potentiate apoptosis induced by gefitinib, suggesting OCT4 may contribute to gefitinib resistance in NSCLC.