Merck
  • Fungal endophytic communities associated to the phyllosphere of grapevine cultivars under different types of management.
Fungal endophytic communities associated to the phyllosphere of grapevine cultivars under different types of management.
Fungal biology (2016-11-29)
Carla Marisa Reis Varanda, Mônica Oliveira, Patrick Materatski, Miguel Landum, Maria Ivone Esteves Clara, Maria do Rosário Félix
ABSTRACT
Fungal endophytes present in different asymptomatic grapevine plants (Vitis vinifera L.) located in different vineyards within Alentejo, a highly important viticulture region in Portugal, were identified in this study. Sampled grapevine plants included the three most representative cultivars in the region, Syrah, Cabernet Sauvignon, and Aragonez, growing under two different modes of management, conventional and biological. Sixteen fungal taxa were identified through sequencing of the internal transcribed spacer region. Total number of endophytic fungi isolated showed significant differences both in management mode and in cultivars, with higher numbers in grapevines under conventional mode and from Syrah cultivar. The composition of fungal endophytic communities did not show significant differences among cultivars, but differences were observed between fungal communities isolated from grapevines under biological or conventional modes. The most fungal taxa isolated from grapevines cultivated under biological mode were Alternaria alternata, Cladosporium sp., and Nigrospora oryzae, and under conventional mode Botrytis cinerea, Epicoccum nigrum, and Epicoccum sp. These differences suggest that the different products used in grapevine production have impacts in fungal endophytic composition. Further investigation of the identified fungi with respect to their antagonistic characteristics and potential use in plant protection to ensure food safety is now in course.