Merck
  • Erythrocyte copper chaperone for superoxide dismutase and superoxide dismutase as biomarkers for hepatic copper concentrations in Labrador retrievers.
Erythrocyte copper chaperone for superoxide dismutase and superoxide dismutase as biomarkers for hepatic copper concentrations in Labrador retrievers.
Veterinary journal (London, England : 1997) (2016-12-13)
K Dirksen, Y S Roelen, M E van Wolferen, H S Kruitwagen, L C Penning, I A Burgener, B Spee, H Fieten
ABSTRACT
Hereditary hepatic copper accumulation in Labrador retrievers leads to hepatitis with fibrosis and eventually cirrhosis. The development of a non-invasive blood-based biomarker for copper status in dogs could be helpful in identifying dogs at risk and to monitor copper concentrations during treatment. In this study, two cellular copper metabolism proteins, Cu/Zn superoxide dismutase (SOD1) and its chaperone (copper chaperone for SOD1, CCS) were measured in erythrocytes and tested for association with hepatic copper concentrations in 15 Labrador retrievers with normal or increased hepatic copper concentrations. Antibodies against CCS and SOD1 were applicable for use in canine specimens. This was demonstrated by the loss of immune-reactive bands for CCS and SOD1 in siRNA treated canine bile duct epithelial cells. Erythrocyte CCS and CCS/SOD1 ratios were decreased 2.37 (P <0.001) and 3.29 (P <0.001) fold in the high copper group compared to the normal copper group. Erythrocyte CCS and CCS/SOD1 ratio are potential new biomarkers for hepatic copper concentrations in Labrador retrievers and could facilitate early diagnosis and treatment monitoring for copper-associated hepatitis in dogs.