Merck
  • Clinical implications of APEX1 and Jagged1 as chemoresistance factors in biliary tract cancer.
Clinical implications of APEX1 and Jagged1 as chemoresistance factors in biliary tract cancer.
Annals of surgical treatment and research (2017-01-17)
Hong-Beum Kim, Won Jin Cho, Nam Gyu Choi, Sung-Soo Kim, Jun Hee Park, Hee-Jeong Lee, Sang Gon Park
ABSTRACT
Biliary cancer is a highly malignant neoplasm with poor prognosis and most patients need to undergo palliative chemotherapy, however major clinical problem associated with the use of chemotherapy is chemoresistance. So far, we aimed at investigating clinical implications of apurinic/apyrimidinic endodeoxyribonuclease 1 (APEX1) and Jagged1 as chemoresistance factors in biliary tract cancer. We used 5 human biliary tract cancer cell lines (SNU-245, SNU-308, SNU-478, SNU-1079, and SNU-1196), and investigated the chemosensitivity of APEX1 and Jagged1 through 3-(4, 5-dimethylthiazol-2-yl)-2, 5-diphenyltetrazolium bromide (MTT) assay and Western blot. Alternately, the 10 patients of advanced biliary cancer consist of 2 group according to the chemotherapy response examined by immunohistochemistry using APEX1 and Jagged1 antibody, and protein expression level was scored for staining intensity and percent positive cell. The result of MTT assay after APEX1 knockdown showed that strong coexpression of APEX1 and Jagged1 cell line (SNU-245, SNU-1079, and SNU-1196) showed a greater decrease in IC These results demonstrate that simultaneous high expression of APEX1 and Jagged1 is associated with chemoresistance in biliary cancer and suggest that is a potential therapeutic target for chemoresistance in advanced biliary cancer.