Merck
  • A rapid streptavidin-capture ELISA specific for the detection of antibodies to feline foamy virus.
A rapid streptavidin-capture ELISA specific for the detection of antibodies to feline foamy virus.
Journal of Immunological Methods (1997)
Winkler IG et al