- Inhibition of L-Type Ca
Inhibition of L-Type Ca
The Journal of neuroscience : the official journal of the Society for Neuroscience (2017-03-05)
Yuyang Sun, Haopeng Zhang, Senthil Selvaraj, Pramod Sukumaran, Saobo Lei, Lutz Birnbaumer, Brij B Singh
PMID28258168
ABSTRACT
Loss of dopaminergic (DA) neurons leads to Parkinson's disease; however, the mechanism(s) for the vulnerability of DA neurons is(are) not fully understood. We demonstrate that TRPC1 regulates the L-type Ca