Merck
  • T-bet-mediated Tim-3 expression dampens monocyte function during chronic hepatitis C virus infection.
T-bet-mediated Tim-3 expression dampens monocyte function during chronic hepatitis C virus infection.
Immunology (2016-11-04)
Wenjing Yi, Peixin Zhang, Yan Liang, Yun Zhou, Huanjun Shen, Chao Fan, Jonathan P Moorman, Zhi Q Yao, Zhansheng Jia, Ying Zhang
ABSTRACT
Hepatitis C virus (HCV) induces a high rate of chronic infection via dysregulation of host immunity. We have previously shown that T-cell immunoglobulin and mucin domain protein-3 (Tim-3) is up-regulated on monocyte/macrophages (M/Mφ) during chronic HCV infection; little is known, however, about the transcription factor that controls its expression in these cells. In this study, we investigated the role of transcription factor, T-box expressed in T cells (T-bet), in Tim-3 expression in M/Mφ in the setting of HCV infection. We demonstrate that T-bet is constitutively expressed in resting CD14