Merck
  • Graphitic carbon coupled poly(anthraquinone) for proton shuttle flow-in-a-cell application.
Graphitic carbon coupled poly(anthraquinone) for proton shuttle flow-in-a-cell application.
Physical chemistry chemical physics : PCCP (2017-03-14)
Selvam Mathi, Rudra Kumar, Rajaram K Nagarale, Ashutosh Sharma
ABSTRACT
Coupled electron and proton transport are an integral part of non-gassing electro-osmotic pumps (EOP). The kinetics of the electrode limits the kinetics of the electron transfer and hence the flow. This is observed in the present study with newly synthesized graphitic carbon covalently coupled to poly(anthraquinone) (PAQ). When EOP with identical electrodes were assembled, proton shuttle maintained the reversible flow, which was linearly dependent on the k