Merck
  • Aldosterone activates the oncogenic signals ERK1/2 and STAT3 via redox-regulated mechanisms.
Aldosterone activates the oncogenic signals ERK1/2 and STAT3 via redox-regulated mechanisms.
Molecular carcinogenesis (2017-03-09)
Nina Queisser, Nicole Schupp, Eva Schwarz, Christina Hartmann, Gerardo G Mackenzie, Patricia I Oteiza
ABSTRACT
Epidemiological studies found an increased risk for kidney cancer in hypertensive patients, of which a subgroup has high aldosterone (Ald) levels. We recently showed that Ald is genotoxic both in kidney tubular cells and in rats with mineralocorticoid-mediated hypertension. The present work investigated in vitro and in vivo, if the oxidative stress-mediated activation of the ERK1/2 pathway, and its downstream target STAT3, could be one mechanism involved in the potential oncogenic capability of excess Ald exposure. The effects of excess Ald were investigated in LLC-PK1 cells and in Ald-induced hypertensive rats. Ald caused cRaf, MEK1/2, and ERK1/2 phosphorylation both in LLC-PK1 cells and in rat kidneys. ERK1/2 activation led to an increased phosphorylation of MSK1, p90RSK, and STAT3. The involvement of ERK1/2 in the activation of STAT3 was evidenced by the capacity of the MEK inhibitor U0126 to prevent Ald-mediated ERK1/2 and STAT3 phosphorylation. Both in vitro and in vivo, the activation of ERK1/2 and STAT3 by Ald was dependent on the mineralocorticoid receptor and was triggered by an increase in cellular oxidants. Ald-mediated oxidant increase was in part due to the activation of the enzymes NADPH oxidase and NO synthase. Proliferation was significantly enhanced and apoptosis decreased in Ald-treated rat kidneys and/or LLC-PK1 cells. Results support the concept that the oxidant-mediated long-term activation of ERK1/2/STAT3 by persistently high Ald levels could trigger proliferative and prosurvival events. Ald-mediated promotion of cell survival and DNA damage could result in kidney cell transformation and initiation of cancer in hypertensive patients with hyperaldosteronism.
Sigma-Aldrich
Anti-PCNA Antibody, clone PC10, clone PC10, Chemicon®, from mouse