- Enrofloxacin and Toltrazuril Are Able to Reduce
Enrofloxacin and Toltrazuril Are Able to Reduce
Frontiers in cellular and infection microbiology (2017-08-12)
Rafaela J da Silva, Angelica O Gomes, Priscila S Franco, Ariane S Pereira, Iliana C B Milian, Mayara Ribeiro, Paolo Fiorenzani, Maria C Dos Santos, José R Mineo, Neide M da Silva, Eloisa A V Ferro, Bellisa de Freitas Barbosa
PMID28798905
ABSTRACT
Classical treatment for congenital toxoplasmosis is based on combination of sulfadiazine and pyrimethamine plus folinic acid. Due to teratogenic effects and bone marrow suppression caused by pyrimethamine, the establishment of new therapeutic strategies is indispensable to minimize the side effects and improve the control of infection. Previous studies demonstrated that enrofloxacin and toltrazuril reduced the incidence of