Merck
  • Revisiting the substrate specificity of mammalian α1,6-fucosyltransferase reveals that it catalyzes core fucosylation of
Revisiting the substrate specificity of mammalian α1,6-fucosyltransferase reveals that it catalyzes core fucosylation of
The Journal of biological chemistry (2017-07-22)
Qiang Yang, Roushu Zhang, Hui Cai, Lai-Xi Wang
ABSTRACT
The mammalian α1,6-fucosyltransferase (FUT8) catalyzes the core fucosylation of