- Tris (1,3-dichloro-2-propyl) phosphate induces toxicity by stimulating CaMK2 in PC12 cells.
Tris (1,3-dichloro-2-propyl) phosphate induces toxicity by stimulating CaMK2 in PC12 cells.
Environmental toxicology (2017-02-10)
Chaonan Li, Li Li, Bencheng Lin, Yanjun Fang, Honglian Yang, Huanliang Liu, Zhuge Xi
PMID28181390
ABSTRACT
Tris (1,3-dichloro-2-propyl) phosphate (TDCIPP) is one of the widely used organophosphorus flame retardants (OPFRs), which are regarded as suitable substitutes for brominated flame retardants (BFRs). Previously, we have validated the toxicity of TDCIPP in PC12 cells owing to the induced alterations in GAP43, NF-H, CaMK2a/2b, and tubulin α/β proteins; however, limited information is currently available on the toxicity and mechanism of TDCIPP. In the present study, cytotoxicity effects were evaluated by exposing PC12 cells to different concentrations of TDCIPP (0-50 μM) for 4 days. To explore the possible mechanisms through which cytotoxicity is induced, changes in intracellular [Ca