Merck
  • miR-133b Inhibits Cell Growth, Migration, and Invasion by Targeting MMP9 in Non-Small Cell Lung Cancer.
miR-133b Inhibits Cell Growth, Migration, and Invasion by Targeting MMP9 in Non-Small Cell Lung Cancer.
Oncology research (2016-12-13)
Yan Zhen, Jia Liu, Yujie Huang, Yajun Wang, Wen Li, Jun Wu
ABSTRACT
Although increasing evidence indicates that the deregulation of microRNAs (miRNAs) contributes to tumorigenesis and invasion, little is known about the role of miR-133b in human non-small cell lung cancer (NSCLC). In the present study, we revealed that the introduction of miR-133b dramatically suppressed NSCLC cell growth, migration, and invasion in vitro. On the contrary, miR-133b inhibitors promoted cell growth, migration, and invasion in vitro. Further studies revealed that matrix metallopeptidase 9 (MMP9) is a direct target gene of miR-133b. Silencing MMP9 inhibited cell growth, migration, and invasion of NSCLC cells, which was consistent with the effect of miR-133b overexpression. In clinical specimens, reduced miR-133b was an unfavorable factor and negatively correlated with MMP9 expression. Our studies demonstrate that miR-133b inhibits cell growth, migration, and invasion by targeting MMP9 in NSCLC.