Merck
  • Cytochrome P450 3A selectively affects the pharmacokinetic interaction between erlotinib and docetaxel in rats.
Cytochrome P450 3A selectively affects the pharmacokinetic interaction between erlotinib and docetaxel in rats.
Biochemical pharmacology (2017-07-19)
Xuan Qin, Jian Lu, Peili Wang, Peipei Xu, Mingyao Liu, Xin Wang
ABSTRACT
Erlotinib as a first-line drug is used in non-small cell lung cancer (NSCLC) patients with sensitive EGFR mutations, while resistance to this drug will occur after several years of treatment. Therefore, the microtubule disturber docetaxel is introduced as combined regimen in clinical trials. This report investigated the potentials and mechanisms of drug-drug interaction (DDI) between erlotinib and docetaxel using wild type (WT) and Cyp3a1/2 knockout (KO) rats. The erlotinib O-demethylation and docetaxel hydroxylation reactions in the absence or the presence of another drug were analyzed in vitro via the assay of rat liver microsomes. In whole animal studies, erlotinib and docetaxel were given to WT and KO rats individually or jointly, and the pharmacokinetic profiles of these two drugs were analyzed and compared among different groups. The results showed that docetaxel not only inhibited the CYP3A-mediated biotransformation of erlotinib in vitro, but also significantly increased the maximum concentration and systemic exposure of erlotinib in vivo in WT rats. In contrast, the DDI was significantly attenuated in KO rats. On the other hand, erlotinib did not influence docetaxel either in vitro biotransformation or in vivo pharmacokinetic behaviors. These results exhibited the potentials of erlotinib-docetaxel interaction and indicated that the CYP3A played the perpetrating role of docetaxel on erlotinib in rats. A better understanding of this DDI with CYP3A may help the regulation of the use of these two drugs, avoid potential problems, and adjust dose carefully and early in clinic.