Merck
  • ADAMTS1-mediated targeting of TSP-1 by PPARδ suppresses migration and invasion of breast cancer cells.
ADAMTS1-mediated targeting of TSP-1 by PPARδ suppresses migration and invasion of breast cancer cells.
Oncotarget (2017-12-08)
Sun Ah Ham, Taesik Yoo, Won Jin Lee, Jung Seok Hwang, Jinwoo Hur, Kyung Shin Paek, Dae-Seog Lim, Sung Gu Han, Chi-Ho Lee, Han Geuk Seo
ABSTRACT
Migration and invasion of cancer cells into surrounding tissue is a key stage of cancer metastasis. Here, we show that peroxisome proliferator-activated receptor (PPAR) δ regulates migration and invasion of human breast cancer cells via thrombospondin-1 (TSP-1) and its degrading protease, a disintegrin and metalloprotease domains with thrombospondin motifs 1 (ADAMTS1). Activation of PPARδ by GW501516, a specific ligand for PPARδ, led to marked inhibition in the cell migration and TSP-1 expression of breast cancer. These effects were suppressed by small interfering RNA-mediated knock-down of ADAMTS1, indicating that ADAMTS1 is involved in PPARδ-mediated inhibition of migration and TSP-1 expression in breast cancer cells. In addition, ligand-activated PPARδ upregulated expression of ADAMTS1 at the transcriptional level via binding of PPARδ to a direct repeat-1 site within the ADAMTS1 gene promoter. Furthermore, ligand-activated PPARδ suppressed invasion of breast cancer cells in an ADAMTS1-dependent manner. Taken together, these results demonstrate that PPARδ suppresses migration and invasion of breast cancer cells by downregulating TSP-1 in a process mediated by upregulation of ADAMTS1.
Sigma-Aldrich
Mitomycin C from Streptomyces caespitosus, powder, BioReagent, suitable for cell culture