Merck
  • Interleukin-33 attenuates doxorubicin-induced cardiomyocyte apoptosis through suppression of ASK1/JNK signaling pathway.
Interleukin-33 attenuates doxorubicin-induced cardiomyocyte apoptosis through suppression of ASK1/JNK signaling pathway.
Biochemical and biophysical research communications (2017-10-03)
Yongwei Yao, Rong Chen, Chunyang Ying, Guohui Zhang, Tao Rui, Aibin Tao
ABSTRACT
Interleukin-33 (IL-33), a new member of the IL-1 cytokine family, has cardiac protective effect in many circumstances. The aims of present study are to assess whether IL-33 can protect cardiomyocytes from doxorubicin (DOX)-induced apoptosis and the mechanism involved in the protection. Cardiomyocytes derived from either wild-type or c-Jun N-terminal kinase deficient (JNK