Merck
  • LCST Phase Behavior and Complexation with Water of an Ionic Liquid Incorporating the 5-Phenyltetrazolate Anion.
LCST Phase Behavior and Complexation with Water of an Ionic Liquid Incorporating the 5-Phenyltetrazolate Anion.
Chemphyschem : a European journal of chemical physics and physical chemistry (2017-08-30)
Leila Moura, Lucy C Brown, Marijana Blesic, John D Holbrey
ABSTRACT
The use of 5-phenyltetrazole, a bioisostere of benzoic acid, as an anion source to prepare an ionic liquid is described for the first time. Tetrabutylphosphonium 5-phenyltetrazolate undergoes phase separation from water with lower critical solution temperature phase behavior, in contrast to completely water miscible tetrabutylphosphonium benzoate, and also exhibits strong complexation with water with both eutectic and peritectic behavior that has not previously been observed in ionic liquid+water systems. The anhydrous and trihydrate salts were isolated and characterized by single crystal X-ray diffraction.