- Ly6C
Ly6C
Biochemical and biophysical research communications (2017-10-24)
Jung Hwa Ko, Hyun Ju Lee, Hyun Jeong Jeong, Joo Youn Oh
PMID29056505
ABSTRACT
The cells of the innate immune system, in addition to their capacity to elicit immunity, play a substantial role in immune tolerance induction. Our group has recently shown that a distinct subset of MHC II