Merck
  • Methylation-induced downregulation and tumor suppressive role of microRNA-29b in gastric cancer through targeting LASP1.
Methylation-induced downregulation and tumor suppressive role of microRNA-29b in gastric cancer through targeting LASP1.
Oncotarget (2017-12-10)
Hui Li, Guoqing Liu, Ke Pan, Xiongying Miao, Yong Xie
ABSTRACT
MicroRNAs (miRs) have been demonstrated to play promoting or tumor suppressive roles in various human cancers, but the regulatory mechanism of miR-29b underlying gastric cancer development and progression still remains largely unclear. In the present study, we found that miR-29b was significantly downregulated in gastric cancer tissues and cell lines. Low expression of miR-29b was significantly associated with DNA methylation, and treatment with DNA methyltransferase inhibitor 5-Aza-20-deoxycytidine upregulated miR-29b in gastric cancer cells. In addition, both reduced miR-29b expression and miR-29b methylation were associated with disease progression and poor prognosis in gastric cancer. Restoration of miR-29b caused a reduction in gastric cancer cell proliferation, migration, and invasion, and inhibited tumor growth