Merck
Using ZnO-Cr
Physical chemistry chemical physics : PCCP (2017-12-01)
Xiaodong Zhu, Jin-Hao Jhang, Chao Zhou, Omur E Dagdeviren, Zheng Chen, Udo D Schwarz, Eric I Altman
ABSTRACT
The ability to affect the surface properties of non-polar Cr