- The influence of amine structures on the stability and catalytic activity of gold nanoparticles stabilized by amine-modified hyperbranched polymers.
The influence of amine structures on the stability and catalytic activity of gold nanoparticles stabilized by amine-modified hyperbranched polymers.
Nanotechnology (2017-12-13)
Hui Dong, Yu Dai, Xiaojin Zhang, Zhenguo Zhang, Shuangli Fu, Zhenlin Zhong
PMID29231179
ABSTRACT
Amine-modified amphiphilic hyperbranched polymers (MePEG-H104-Nx) were prepared from hyperbranched 2,2-bis(methylol)propionic acid polyester (H104) by decoration with polyethylene glycol monomethyl ether (MePEG) and different classes of oligo(ethylenimine)s. By using the MePEG-H104-Nx polymers as stabilizers, gold nanoparticles (AuNPs) were prepared in an aqueous medium by the reduction of HAuCl