Merck
  • Myeloid receptor CD36 is required for early phagocytosis of myocardial infarcts and induction of Nr4a1-dependent mechanisms of cardiac repair.
Myeloid receptor CD36 is required for early phagocytosis of myocardial infarcts and induction of Nr4a1-dependent mechanisms of cardiac repair.
FASEB journal : official publication of the Federation of American Societies for Experimental Biology (2017-09-02)
Shirley Dehn, Edward B Thorp
ABSTRACT
Phagocytosis after myocardial infarction (MI) is a prerequisite to cardiac repair. Recruited monocytes clear necrotic cardiomyocytes and differentiate into cardiac macrophages. Some studies have linked apoptotic cell receptors on cardiac macrophages to tissue repair; however, the contribution of precursor monocyte phagocytic receptors, which are the first to interact with the cardiac parenchyma, is unclear. The scavenger receptor cluster of differentiation (CD)36 protein was detected on cardiac Ly6cHI monocytes, and bone marrow-derived
Sigma-Aldrich
ChIPAb+ CREB - ChIP Validated Antibody and Primer Set, rabbit monoclonal, culture supernatant, clone NL904, from rabbit