Merck
  • Hypoxia primes human normal prostate epithelial cells and cancer cell lines for the NLRP3 and AIM2 inflammasome activation.
Hypoxia primes human normal prostate epithelial cells and cancer cell lines for the NLRP3 and AIM2 inflammasome activation.
Oncotarget (2016-04-09)
Ravichandran Panchanathan, Hongzhu Liu, Divaker Choubey
ABSTRACT
The molecular mechanisms by which hypoxia contributes to prostatic chronic inflammation (PCI) remain largely unknown. Because hypoxia stimulates the transcriptional activity of NF-κB, which "primes" cells for inflammasome activation by inducing the expression of NLRP3 or AIM2 receptor and pro-IL-1β, we investigated whether hypoxia could activate the NLRP3 and AIM2 inflammasome in human normal prostate epithelial cells (PrECs) and cancer cell lines. Here we report that hypoxia (1% O2) treatment of PrECs, prostate cell lines, and a macrophage cell line (THP-1) increased the levels of NLRP3, AIM2, and pro-IL-1β. Further, hypoxia in cells potentiated activation of the NLRP3 and AIM2 inflammasome activity. Notably, hypoxia "primed" cells for NLRP3 and AIM2 inflammasome activation through stimulation of the NF-κB activity. Our observations support the idea that hypoxia in human prostatic tumors contributes to PCI, in part, by priming cells for the activation of NLRP3 and AIM2 inflammasome.