Merck
  • Control of RAB7 activity and localization through the retromer-TBC1D5 complex enables RAB7-dependent mitophagy.
Control of RAB7 activity and localization through the retromer-TBC1D5 complex enables RAB7-dependent mitophagy.
The EMBO journal (2017-11-22)
Ana Jimenez-Orgaz, Arunas Kvainickas, Heike Nägele, Justin Denner, Stefan Eimer, Jörn Dengjel, Florian Steinberg
ABSTRACT
Retromer is an endosomal multi-protein complex that organizes the endocytic recycling of a vast range of integral membrane proteins. Here, we establish an additional retromer function in controlling the activity and localization of the late endosomal small GTPase RAB7. Surprisingly, we found that RAB7 not only decorates late endosomes or lysosomes, but is also present on the endoplasmic reticulum,