Merck
  • One-way membrane trafficking of SOS in receptor-triggered Ras activation.
One-way membrane trafficking of SOS in receptor-triggered Ras activation.
Nature structural & molecular biology (2016-08-09)
Sune M Christensen, Hsiung-Lin Tu, Jesse E Jun, Steven Alvarez, Meredith G Triplet, Jeffrey S Iwig, Kamlesh K Yadav, Dafna Bar-Sagi, Jeroen P Roose, Jay T Groves
ABSTRACT
SOS is a key activator of the small GTPase Ras. In cells, SOS-Ras signaling is thought to be initiated predominantly by membrane recruitment of SOS via the adaptor Grb2 and balanced by rapidly reversible Grb2-SOS binding kinetics. However, SOS has multiple protein and lipid interactions that provide linkage to the membrane. In reconstituted-membrane experiments, these Grb2-independent interactions were sufficient to retain human SOS on the membrane for many minutes, during which a single SOS molecule could processively activate thousands of Ras molecules. These observations raised questions concerning how receptors maintain control of SOS in cells and how membrane-recruited SOS is ultimately released. We addressed these questions in quantitative assays of reconstituted SOS-deficient chicken B-cell signaling systems combined with single-molecule measurements in supported membranes. These studies revealed an essentially one-way trafficking process in which membrane-recruited SOS remains trapped on the membrane and continuously activates Ras until being actively removed via endocytosis.