Merck
A
Disease models & mechanisms (2017-10-14)
Zachary Fischer, Rahul Das, Anna Shipman, Jin-Yuan Fan, Laramie Pence, Samuel Bouyain, Leonard L Dobens
ABSTRACT
Members of the Tribbles family of proteins are conserved pseudokinases with diverse roles in cell growth and proliferation. Both