Merck
  • Antioxidant Potential and Modulatory Effects of Restructured Lipids from the Amazonian Palms on 
Liver Cells.
Antioxidant Potential and Modulatory Effects of Restructured Lipids from the Amazonian Palms on 
Liver Cells.
Food technology and biotechnology (2018-03-16)
Andrea de Oliveira Falcão, Paula Speranza, Tatiane Ueta, Isabela Mateus Martins, Gabriela Alves Macedo, Juliana Alves Macedo
ABSTRACT
Enzymatic interesterification is used to manipulate oil and fat in order to obtain improved restructured lipids with desired technological properties. However, with raw materials containing significant amounts of bioactive compounds, the influence of this enzymatic process on the bioactivity of the final product is still not clear. Thus, the aim of this study is to evaluate the antioxidant potential and modulatory effects of two raw materials from the Amazonian area, buriti oil and murumuru fat, before and after lipase interesterification, on human hepatoma cells (HepG2). The results indicate that minor bioactive compounds naturally found in the raw materials and their antioxidant capacity are preserved after enzymatic interesterification, and that the restructured lipids modulate HepG2 endogenous antioxidant enzyme.