Merck
  • Androgen Triggers the Pro-Migratory CXCL12/CXCR4 Axis in AR-Positive Breast Cancer Cell Lines: Underlying Mechanism and Possible Implications for the Use of Aromatase Inhibitors in Breast Cancer.
Androgen Triggers the Pro-Migratory CXCL12/CXCR4 Axis in AR-Positive Breast Cancer Cell Lines: Underlying Mechanism and Possible Implications for the Use of Aromatase Inhibitors in Breast Cancer.
Cellular physiology and biochemistry : international journal of experimental cellular physiology, biochemistry, and pharmacology (2017-11-14)
Kalliopi Azariadis, Fotini Kiagiadaki, Vasiliki Pelekanou, Vasiliki Bempi, Kostas Alexakis, Marilena Kampa, Andreas Tsapis, Elias Castanas, George Notas
ABSTRACT
Reports regarding the role of androgen in breast cancer (BC) are conflicting. Some studies suggest that androgen could lead to undesirable responses in the presence of certain BC tumor characteristics. We have shown that androgen induces C-X-C motif chemokine 12 (CXCL12) in BC cell lines. Our aim was to identify the mechanisms regulating the phenotypic effects of androgen-induced CXCL12 on Androgen Receptor (AR) positive BC cell lines. We analyzed the expression of CXCL12 and its receptors with qPCR and ELISA and the role of Nuclear Receptor Coactivator 1 (NCOA1) in this effect. AR effects on the CXCL12 promoter was studied via Chromatin-immunoprecipitation. We also analyzed publically available data from The Cancer Genome Atlas to verify AR-CXCL12 interactions and to identify the effect or Aromatase Inhibitors (AI) therapy on CXCL12 expression and disease progression in AR positive cases. CXCL12 induction occurs only in AR-positive BC cell lines, possibly via an Androgen Response Element, upstream of the CXCL12 promoter. The steroid receptor co-regulator NCOA1 is critical for this effect. Androgen only induced the motility of p53-mutant BC cells T47D cells via upregulation of CXCR4 expression while they had no effect on wild-type p53 MCF-7 cells. Loss of CXCR4 expression and depletion of CXCL12 abolished the effect of androgen in T47D cells while inhibition of p53 expression in MCF-7 cells made them responsive to androgen and increased their motility in the presence to androgen. Patients with estrogen receptor positive (ER+)/AR+ BC treated with AIs were at increased risk of disease progression compared to ER+/AR+ non-AI treated and ER+/AR- AI treated cases. AIs may lead to unfavorable responses in some ER/AR positive BC cases, especially in patients with AR+, p53 mutant tumors.