Skip to Content
Merck

GF93012768

Tungsten

tube, outside diameter 1.6 mm, length 50 mm, wall thickness 0.5 mm

Synonym(s):

W 007200, W

Sign In to View Organizational & Contract Pricing.

Select a Size

1 EA

PLN 3,870.00

PLN 3,870.00


Please contact Customer Service for Availability

Request a Bulk Order

About This Item

Empirical Formula (Hill Notation):
W
CAS Number:
Molecular Weight:
183.84
NACRES:
NA.23
PubChem Substance ID:
UNSPSC Code:
12141747
MDL number:

Skip To

Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist

Product Name

Tungsten, tube, outside diameter 1.6 mm, length 50 mm, wall thickness 0.5 mm

InChI

1S/W

SMILES string

[W]

InChI key

WFKWXMTUELFFGS-UHFFFAOYSA-N

form

tubes

manufacturer/tradename

Goodfellow 930-127-68

resistivity

4.9 μΩ-cm, 20°C

L

50 mm

O.D.

1.6 mm

wall thickness

0.5 mm

bp

5660 °C (lit.)

mp

3410 °C (lit.)

density

19.3 g/mL at 25 °C (lit.)

Compare Similar Items

View Full Comparison

Show Differences

1 of 4

This Item
GF40617121GF13753605GF72275339
manufacturer/tradename

Goodfellow 930-127-68

manufacturer/tradename

Goodfellow 406-171-21

manufacturer/tradename

Goodfellow 137-536-05

manufacturer/tradename

Goodfellow 722-753-39

form

tubes

form

foil

form

wire

form

wire

resistivity

4.9 μΩ-cm, 20°C

resistivity

4.9 μΩ-cm, 20°C

resistivity

4.9 μΩ-cm, 20°C

resistivity

4.9 μΩ-cm, 20°C

density

19.3 g/mL at 25 °C (lit.)

density

19.3 g/mL at 25 °C (lit.)

density

19.3 g/mL at 25 °C (lit.)

density

19.3 g/mL at 25 °C (lit.)

bp

5660 °C (lit.)

bp

5660 °C (lit.)

bp

5660 °C (lit.)

bp

5660 °C (lit.)

mp

3410 °C (lit.)

mp

3410 °C (lit.)

mp

3410 °C (lit.)

mp

3410 °C (lit.)

General description

For updated SDS information please visit www.goodfellow.com

Legal Information

Product of Goodfellow

Storage Class

11 - Combustible Solids

wgk

nwg

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Carina Coimbra et al.
Systematic and applied microbiology, 40(6), 388-395 (2017-08-15)
Tungsten (W) has industrial and economic importance, and is in the European Union list of metals with a high supply risk. It is used by living organisms, which transport it into the cell, in the form of tungstate ion (WO

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service