Skip to Content
Merck

BDPH9BCDA653

Bide Pharmatech

Pyrido[2,3-d]pyrimidin-4(1H)-one

Synonym(s):

pyrido[2,3-d]pyrimidin-4(3H)-one

Sign In to View Organizational & Contract Pricing.

Select a Size


About This Item

Linear Formula:
C7H5O1N3
CAS Number:
Molecular Weight:
147.14
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist

Inchi Code

1S/C7H5N3O/c11-7-5-2-1-3-8-6(5)9-4-10-7/h1-4H,(H,8,9,10,11)

Inchi Key

XKEBMWRWBWRQAO-UHFFFAOYSA-N

Shipping Temperature

Normal

Purity

97%

Storage temp.

Sealed in dry, room temperature

Country of Origin

CN

Physical Form

White to beige to brown powder or crystals

Looking for similar products? Visit Product Comparison Guide

signalword

Warning

pictograms

Exclamation mark

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service