grade
certified reference material
agency
National Research Council of Canada (NRC)
description
Analyte group-Total element content
manufacturer/tradename
National Research Council of Canada (NRC)
application(s)
food and beverages
format
matrix material
storage temp.
2-8°C
General description
CASS-6 is a nearshore seawater Certified Reference Material (CRM) prepared at the National Research Council of Canada (NRC) with information on trace element content and other constituents. A unit of CASS-6 consists of approximately 200 mL of seawater. This material is intended for use in the method development, validation, and quality control for the analysis of trace and matrix constituents in seawater matrices. For more information, please refer to the Certificate of Analysis or Safety Data Sheet https://doi.org/10.4224/crm.2016.cass-6
Features and Benefits
NRC Canada’s certified reference materials (CRMs) are traceable to the International System of Units, and conform to the requirements of ISO/IEC 17025 and ISO 17034. For specific values, as well as additional information regarding stability and storage information, please view the Certificate of Analysis. With NRC Canada’s CRMs, you gain access to products backed by extensive research and expertise, fostering trust and consistency in your results. Elevate your laboratory’s accuracy and credibility with NRC Canada’s certified reference materials.
Analysis Note
These products are for test and assay use only. They are not meant for administration to humans or animals and cannot be used to diagnose, treat, or cure diseases of any kind.
Other Notes
Tested for the analytes listed below. See certificate for values and more details
Trace Elements: Boron (B), Cadmium (Cd), Cobalt (Co), Copper (Cu), Iron (Fe), Lead (Pb), Manganese (Mn), Molybdenum (Mb), Nickel (Ni), Uranium (U), Zinc (Zn)
Trace Elements: Boron (B), Cadmium (Cd), Cobalt (Co), Copper (Cu), Iron (Fe), Lead (Pb), Manganese (Mn), Molybdenum (Mb), Nickel (Ni), Uranium (U), Zinc (Zn)
Sales restrictions may apply.
Storage Class
12 - Non Combustible Liquids
wgk
nwg
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.