Skip to Content
Merck

CRM-AZA2-B

Azaspiracid-2 Calibration Solution CRM

certified reference material

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

UNSPSC Code:
41116107
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


grade

certified reference material

agency

National Research Council of Canada (NRC)

description

Analyte group-Algal toxins

manufacturer/tradename

National Research Council of Canada (NRC)

application(s)

food and beverages

format

single component solution

storage temp.

−70°C

General description

CRM-AZA2-b is designed for use as an instrument calibration solution to aid the analyst in the determination of AZA2 in plankton and shellfish sample extracts. The most widely used method of analysis is liquid chromatography-mass spectrometry (LC-MS) and the concentration of AZA2 in CRM AZA2-b is suitable for the calibration of most LC-MS instruments. CRM-AZA2-b can also be used for spiking shellfish control samples for recovery experiments. For more information, please refer to the Certificate of Analysis or Safety Data Sheet.

Features and Benefits

NRC Canada’s certified reference materials (CRMs) are traceable to the International System of Units, and conform to the requirements of ISO/IEC 17025 and ISO 17034. For specific values, as well as additional information regarding stability and storage information, please view the Certificate of Analysis. With NRC Canada’s CRMs, you gain access to products backed by extensive research and expertise, fostering trust and consistency in your results. Elevate your laboratory’s accuracy and credibility with NRC Canada’s certified reference materials.

Analysis Note

These products are for test and assay use only. They are not meant for administration to humans or animals and cannot be used to diagnose, treat, or cure diseases of any kind.

Other Notes

Sales restrictions may apply.


signalword

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Flam. Liq. 2 - STOT SE 1

target_organs

Eyes,Central nervous system

Storage Class

3 - Flammable liquids

wgk

WGK 2

flash_point_f

49.5 °F - closed cup

flash_point_c

9.7 °C - closed cup


Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

67-56-1

CAS No.


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Global Trade Item Number

SKUGTIN
CRM-AZA2-B04065270773074