Product Name
(R)-(+)-1-(1-Naphthyl)ethylamine derivatization grade (chiral), LiChropur, ≥99.5%
Product Number
70710
Product Brand
sial
CAS Number
3886-70-2
Molecular Weight
171.24
Test
Specification
APPEARANCE (COLOR)Colorless to Light Yellow
APPEARANCE (FORM)Liquid
PURITY (GC AREA %)≥ 99.5 %
ENANTIOMERIC RATIO≥ 99.5%: 0.5%
SPECIFIC ROTATION (20/D)57 ± 2 Degrees
CONCENTRATIONC=2 IN ETHANOL
REFRACTIVE INDEX N20/D1.623 - 1.625
PROTON NMR SPECTRUMCONFORMS TO STRUCTURE
RECOMMENDED RETEST PERIOD42 MONTHS