Product Name
Sodium taurocholate hydrate ≥97.0% (TLC)
Product Number
86339
Product Brand
sigma
CAS Number
345909-26-4
Molecular Weight
537.68 (anhydrous basis)
Test
Specification
APPEARANCE (COLOR)Colorless or White
APPEARANCE (FORM)Powder
THIN LAYER CHROMATOGR.≥ 97.0 % TLC
REM. ON CHROMATOGRAPHYCholic acid < 2 %; Taurin < 1 %
SPECIFIC ROTATION (20/D)24 ± 1 Degrees
CONCENTRATIONC=3 IN WATER
REM. ON OPT. ROTATIONIN DRY SUBSTANCE
SOLUBILITY (COLOR)Colorless
SOLUBILITY (TURBIDITY)Clear
SOLUBILITY (METHOD)0.25G IN 5ML WATER
WATER≤ 7.0 %
INFRARED SPECTRUMCONFORMS TO STRUCTURE