Product Name
TrueGel3D Hydrogel Kits CD cell-degradable crosslinker and RGD peptide
Product Number
true1
Product Brand
sigma
CAS Number
Molecular Weight
Test
Specification
TRU-RGD:Hydrogel FormationForms hydrogel within 15 mins
TRU-CD:Thiol groups20-24 mmol/L
TRU-REC: pHpH 7.4-7.5 at 20-25 °C