Merck
3-[(3-Cholamidopropyl)dimethylammonio]-1-propanesulfonate hydrate 98%

3-[(3-Cholamidopropyl)dimethylammonio]-1-propanesulfonate hydrate

3-[(3-Cholamidopropyl)dimethylammonio]-1-propanesulfonate hydrate 98%
Synonyms:
CHAPS
Empirical Formula (Hill Notation):
C32H58N2O7S · xH2O
CAS Number:
Molecular Weight:
614.88 (anhydrous basis)

Product Comparison Guide

Use the product attributes below to configure the comparison table. (Select up to 3 total.)

Select Attribute
Select Attribute
Select Attribute
Sort by: Default
Product Number
Product Description
Pricing
3-[(3-Cholamidopropyl)dimethylammonio]-1-propanesulfonate hydrate, 98%