Skip to Content
Merck
Blocking Reagent For nucleic acid hybridization and detection

Blocking Reagent

Blocking Reagent For nucleic acid hybridization and detection
Synonym(s):
blocking reagent

Products

Product Category
Manufacturer
Physical Form
Technique
Showing 1-2 of 2
Compare
Product Number
Description
Pricing