Skip to Content
Products
Cart
0
PL
EN
Products
Products
Applications
Services
Resources
Support
Login / Register
Order Lookup
Quick Order
Cart
0
Back
Gram’s iodine solution
Synonym(s):
Potassium Triiodide
Products
Product Category
histology special stains
stains
Brand
Sigma-Aldrich
Color
Physical Form
Product Line
Quality Segment
Application
Technique
Filter Products
Showing 1-3 of 3
Compare
Product Number
Description
Pricing
90107
Gram’s iodine solution,
suitable for microscopy
Expand
View Pricing
HT90232
Gram’s iodine solution,
Expand
View Pricing
HT902
Gram’s iodine solution,
Expand
View Pricing
An unknown error has occured.