Skip to Content
Merck
Polyimide (PI), film, thickness 0.125 mm, size 300 × 300 mm, grade kapton hn

Polyimide

Polyimide (PI), film, thickness 0.125 mm, size 300 × 300 mm, grade kapton hn
Synonym(s):
IM301450

Products

Product Category
Manufacturer
Physical Form
Diameter (mm)
Showing 1-20 of 30
Page 1 of 2
Compare
Product Number
Description
Pricing
Page 1 of 2