Skip to Content
Products
Cart
0
PL
EN
Products
Products
Applications
Services
Resources
Support
Login / Register
Order Lookup
Quick Order
Cart
0
Back
Schiff′s reagent
Products
Product Category
salts
stains
Brand
Sigma-Aldrich
Physical Form
Quality Segment
Application
Filter Products
Showing 1-2 of 2
Compare
Product Number
Description
Pricing
1.09033
Schiff′s reagent,
for microscopy
Expand
View Pricing
3952016
Schiff′s reagent,
Expand
View Pricing
An unknown error has occured.