Skip to Content
Merck
Scriptaid ≥95%, solid

Scriptaid

Scriptaid ≥95%, solid
Synonym(s):
6-(1,3-Dioxo-1H, 3H-benzo[de]isoquinolin-2-yl)-hexanoic acid hydroxyamide
Empirical Formula (Hill Notation):
C18H18N2O4
CAS Number:
Molecular Weight:
326.35

Products

Product Category
Brand
Biological Source
Physical Form
Purity
Research Area
Quality Segment
Showing 1-1 of 1
Compare
Product Number
Description
Pricing