Merck
Trometamol European Pharmacopoeia (EP) Reference Standard

Trometamol

Trometamol European Pharmacopoeia (EP) Reference Standard
Synonyms:
Tromethamine, Tris
Empirical Formula (Hill Notation):
C4H11NO3
CAS Number:
Molecular Weight:
121.14
EC Number:

Product Comparison Guide

Use the product attributes below to configure the comparison table. (Select up to 3 total.)

Select Attribute
Select Attribute
Select Attribute
Sort by: Default
Product Number
Product Description
Pricing
Trometamol, European Pharmacopoeia (EP) Reference Standard